Assay Details
| Gene Symbol: MDGA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025150963.1 | XP_025006731.1 | 14 - 15 | 3047 | 59 | |
| XM_025150964.1 | XP_025006732.1 | 13 - 14 | 2999 | 59 | ||
More Information
Set Membership: |
|

MDGA2
Genomic Map