Assay Details
| Gene Symbol: MTERF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004937810.3 | XP_004937867.2 | 2 | 1607 | 90 | |
| XM_004937811.2 | XP_004937868.1 | 2 | 1698 | 90 | ||
| XM_025155620.1 | XP_025011388.1 | 2 | 1989 | 90 | ||
| XM_025155622.1 | XP_025011390.1 | 2 | 1775 | 90 | ||
More Information
Set Membership: |
|

MTERF2
Genomic Map