Assay Details
| Gene Symbol: DSG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025147912.1 | XP_025003680.1 | 14 | 4812 | 77 | |
| XM_426083.6 | XP_426083.4 | 15 | 5042 | 77 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DSG2| Gene Symbol: DSG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025147912.1 | XP_025003680.1 | 14 | 4812 | 77 | |
| XM_426083.6 | XP_426083.4 | 15 | 5042 | 77 | ||
Set Membership: |
|