Assay Details
| Gene Symbol: TMEM61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004936834.3 | XP_004936891.1 | 2 - 3 | 1582 | 109 | |
| XM_025153175.1 | XP_025008943.1 | 2 - 3 | 555 | 109 | ||
| XM_025153176.1 | XP_025008944.1 | 2 - 3 | 514 | 109 | ||
| XM_422496.6 | XP_422496.4 | 2 - 3 | 556 | 109 | ||
More Information
Set Membership: |
|

TMEM61
Genomic Map