Assay Details
| Gene Symbol: MRPL54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003642837.4 | XP_003642885.1 | 2 - 3 | 231 | 147 | |
| GenBank mRNA | BX931282.2 | - | 2 - 3 | 253 | 147 | |
| BX934325.2 | - | 2 - 3 | 232 | 147 | ||
More Information
Set Membership: |
|

MRPL54
Genomic Map