Assay Details
| Gene Symbol: SH3BP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004942501.3 | XP_004942558.1 | 6 | 5414 | 84 | |
| XM_421882.6 | XP_421882.3 | 5 | 5283 | 84 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SH3BP4| Gene Symbol: SH3BP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004942501.3 | XP_004942558.1 | 6 | 5414 | 84 | |
| XM_421882.6 | XP_421882.3 | 5 | 5283 | 84 | ||
Set Membership: |
|