Assay Details
| Gene Symbol: MORN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003642178.4 | XP_003642226.1 | 2 - 3 | 347 | 90 | |
| XM_015275587.2 | XP_015131073.1 | 2 - 3 | 387 | 90 | ||
| XM_015275588.2 | XP_015131074.1 | 2 - 3 | 559 | 90 | ||
| XM_015275589.2 | XP_015131075.1 | 2 - 3 | 291 | 90 | ||
| XM_015275590.2 | XP_015131076.1 | 2 - 3 | 347 | 90 | ||
| XM_015275592.2 | XP_015131078.1 | 2 - 3 | 335 | 90 | ||
More Information
Set Membership: |
|

MORN3
Genomic Map