Assay Details
| Gene Symbol: CFAP126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025143655.1 | XP_024999423.1 | 3 - 4 | 183 | 75 | |
| XM_025143656.1 | XP_024999424.1 | 3 - 4 | 654 | 75 | ||
More Information
Set Membership: |
|

CFAP126
Genomic Map