Assay Details
| Gene Symbol: AKAP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001234243.5 | XP_001234244.1 | 4 - 5 | 552 | 103 | |
| XM_015278632.1 | XP_015134118.1 | 4 - 5 | 2202 | 103 | ||
More Information
Set Membership: |
|

AKAP14
Genomic Map