Assay Details
| Gene Symbol: MMRN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025151897.1 | XP_025007665.1 | 6 | 2805 | 93 | |
| XM_421496.6 | XP_421496.3 | 7 | 2980 | 93 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MMRN2| Gene Symbol: MMRN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025151897.1 | XP_025007665.1 | 6 | 2805 | 93 | |
| XM_421496.6 | XP_421496.3 | 7 | 2980 | 93 | ||
Set Membership: |
|