Assay Details
| Gene Symbol: BPIFB6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015296596.2 | XP_015152082.1 | 13 - 14 | 1643 | 57 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BPIFB6| Gene Symbol: BPIFB6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015296596.2 | XP_015152082.1 | 13 - 14 | 1643 | 57 | |
Set Membership: |
|