Assay Details
| Gene Symbol: FAM222A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015275505.2 | XP_015130991.2 | 3 | 2905 | 94 | |
| XM_025155592.1 | XP_025011360.1 | 2 | 5413 | 94 | ||
| XM_025155593.1 | XP_025011361.1 | 3 | 4707 | 94 | ||
More Information
Set Membership: |
|

FAM222A
Genomic Map