Assay Details
| Gene Symbol: EMX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001232150.5 | XP_001232151.3 | 4 | 989 | 68 | |
| GenBank mRNA | JF297586.1 | - | 1 | 126 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EMX1| Gene Symbol: EMX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001232150.5 | XP_001232151.3 | 4 | 989 | 68 | |
| GenBank mRNA | JF297586.1 | - | 1 | 126 | 68 | |
Set Membership: |
|