Assay Details
| Gene Symbol: TMEM174 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004937367.3 | XP_004937424.1 | 3 | 938 | 88 | |
| XM_015277660.2 | XP_015133146.1 | 3 | 931 | 88 | ||
| XM_025144761.1 | XP_025000529.1 | 3 | 893 | 88 | ||
| GenBank mRNA | CR386164.1 | - | 1 | 422 | 88 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_025144761.1 transcripts.
Set Membership: |
|

TMEM174
Genomic Map