Assay Details
| Gene Symbol: XKRX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004940698.2 | XP_004940755.1 | 5 | 1903 | 73 | |
| XM_004940699.3 | XP_004940756.1 | 5 | 1834 | 73 | ||
| XM_015278491.2 | XP_015133977.1 | 5 | 4490 | 73 | ||
| XM_015278492.2 | XP_015133978.1 | 4 | 1697 | 73 | ||
| XM_025150508.1 | XP_025006276.1 | 4 | 4569 | 73 | ||
| XM_025150509.1 | XP_025006277.1 | 5 | 4133 | 73 | ||
More Information
Set Membership: |
|

XKRX
Genomic Map