Assay Details
| Gene Symbol: USP44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015283925.2 | XP_015139411.1 | 2 | 457 | 100 | |
| XM_015283938.2 | XP_015139424.1 | 4 | 1204 | 100 | ||
| XM_015283945.2 | XP_015139431.1 | 3 | 1097 | 100 | ||
| XM_015283952.2 | XP_015139438.1 | 2 | 1029 | 100 | ||
| XM_025154835.1 | XP_025010603.1 | 2 | 2043 | 100 | ||
| XM_025154841.1 | XP_025010609.1 | 5 | 1656 | 100 | ||
| XM_025154846.1 | XP_025010614.1 | 3 | 2151 | 100 | ||
| XM_025154847.1 | XP_025010615.1 | 3 | 2151 | 100 | ||
| GenBank mRNA | BX931266.1 | - | 2 | 508 | 100 | |
| BX934257.1 | - | 1 | 396 | 100 | ||
More Information
Set Membership: |
|

USP44
Genomic Map