Assay Details
| Gene Symbol: FAM83B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015284902.2 | XP_015140388.1 | 3 - 4 | 1176 | 99 | |
| XM_426203.6 | XP_426203.2 | 3 - 4 | 3802 | 99 | ||
More Information
Set Membership: |
|

FAM83B
Genomic Map