Assay Details
| Gene Symbol: TGM6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001234441.4 | XP_001234442.3 | 12 - 13 | 3572 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TGM6| Gene Symbol: TGM6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001234441.4 | XP_001234442.3 | 12 - 13 | 3572 | 65 | |
Set Membership: |
|