Assay Details
| Gene Symbol: SLC22A31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015292672.2 | XP_015148158.1 | 6 - 7 | 962 | 92 | |
| XM_025154316.1 | XP_025010084.1 | 5 - 6 | 810 | 92 | ||
More Information
Set Membership: |
|

SLC22A31
Genomic Map