Assay Details
| Gene Symbol: LRRC15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025153728.1 | XP_025009496.1 | 1 - 2 | 8159 | 105 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_025153728.1 transcripts.
Set Membership: |
|

LRRC15
Genomic Map