Assay Details
| Gene Symbol: ELFN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015294341.2 | XP_015149827.1 | 2 - 3 | 308 | 62 | |
| XM_025155383.1 | XP_025011151.1 | 2 - 3 | 248 | 62 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015294341.2, XM_025155383.1 transcripts.
Set Membership: |
|

ELFN1
Genomic Map