Assay Details
| Gene Symbol: LOC426897 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015300378.2 | XP_015155864.2 | 1 - 2 | 900 | 95 | |
| XM_424505.6 | XP_424505.5 | 1 - 2 | 895 | 95 | ||
More Information
Set Membership: |
|

LOC426897
Genomic Map