Assay Details
| Gene Symbol: CLDN18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003641730.4 | XP_003641778.1 | 2 - 3 | 458 | 67 | |
| XM_426691.5 | XP_426691.4 | 2 - 3 | 459 | 67 | ||
More Information
Set Membership: |
|

CLDN18
Genomic Map