Assay Details
| Gene Symbol: PLA2G4EL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_426427.6 | XP_426427.5 | 13 - 14 | 1462 | 134 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PLA2G4EL3| Gene Symbol: PLA2G4EL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_426427.6 | XP_426427.5 | 13 - 14 | 1462 | 134 | |
Set Membership: |
|