Assay Details
| Gene Symbol: JAML | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015298256.2 | XP_015153742.1 | 5 - 6 | 914 | 92 | |
| XM_015298257.2 | XP_015153743.1 | 5 - 6 | 994 | 92 | ||
| XM_417915.6 | XP_417915.4 | 3 - 4 | 1880 | 92 | ||
More Information
Set Membership: |
|

JAML
Genomic Map