Assay Details
| Gene Symbol: LHFPL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001234276.5 | XP_001234277.4 | 1 - 2 | 1117 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LHFPL4| Gene Symbol: LHFPL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001234276.5 | XP_001234277.4 | 1 - 2 | 1117 | 91 | |
Set Membership: |
|