Assay Details
| Gene Symbol: RNF43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003642379.4 | XP_003642427.3 | 8 - 9 | 4039 | 64 | |
| XM_025142103.1 | XP_024997871.1 | 7 - 8 | 3886 | 64 | ||
More Information
Set Membership: |
|

RNF43
Genomic Map