Assay Details
| Gene Symbol: OPA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015302721.2 | XP_015158207.1 | 1 | 213 | 133 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015302721.2 transcripts.
Set Membership: |
|

OPA3
Genomic Map