Assay Details
Gene Symbol: LOC100791699 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK244196.1 | - | 1 | 124 | 105 | |
AK244221.1 | - | 1 | 124 | 105 | ||
AK244407.1 | - | 1 | 124 | 105 | ||
AK285600.1 | - | 1 | 124 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
