Assay Details
| Gene Symbol: Znf330 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004868893.2 | XP_004868950.1 | 5 - 6 | 493 | 84 | |
| XM_021247707.1 | XP_021103366.1 | 5 - 6 | 478 | 84 | ||
| XM_021247708.1 | XP_021103367.1 | 5 - 6 | 426 | 84 | ||
More Information
Set Membership: |
|

Znf330
Genomic Map