Assay Details
| Gene Symbol: Mospd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004860101.2 | XP_004860158.1 | 3 - 4 | 498 | 68 | |
| XM_013077909.2 | XP_012933363.1 | 5 - 6 | 683 | 68 | ||
| XM_013077910.2 | XP_012933364.1 | 3 - 4 | 467 | 68 | ||
| XM_013077912.2 | XP_012933366.1 | 3 - 4 | 498 | 68 | ||
| XM_021240596.1 | XP_021096255.1 | 5 - 6 | 643 | 68 | ||
| XM_021240597.1 | XP_021096256.1 | 4 - 5 | 601 | 68 | ||
| XM_021240598.1 | XP_021096257.1 | 4 - 5 | 561 | 68 | ||
More Information
Set Membership: |
|

Mospd1
Genomic Map