Assay Details
| Gene Symbol: Znf711 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004868877.3 | XP_004868934.1 | 4 - 5 | 683 | 78 | |
| XM_004868878.3 | XP_004868935.1 | 2 - 3 | 288 | 78 | ||
| XM_004868879.2 | XP_004868936.1 | 4 - 5 | 678 | 78 | ||
| XM_004868880.2 | XP_004868937.1 | 4 - 5 | 512 | 78 | ||
| XM_013067110.2 | XP_012922564.1 | 4 - 5 | 678 | 78 | ||
| XM_021247652.1 | XP_021103311.1 | 4 - 5 | 567 | 78 | ||
| XM_021247653.1 | XP_021103312.1 | 3 - 4 | 423 | 78 | ||
More Information
Set Membership: |
|

Znf711
Genomic Map