Assay Details
| Gene Symbol: Fth1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001310304.1 | NP_001297233.1 | 3 - 4 | 424 | 61 | |
| GenBank mRNA | KM486707.1 | - | 3 - 4 | 424 | 61 | |
More Information
Set Membership: |
|

Fth1
Genomic Map