Assay Details
| Gene Symbol: Mb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001310236.1 | NP_001297165.1 | 2 - 3 | 373 | 62 | |
| GenBank mRNA | KM486737.1 | - | 2 - 3 | 373 | 62 | |
More Information
Set Membership: |
|

Mb
Genomic Map