Assay Details
| Gene Symbol: Prelid1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004836277.3 | XP_004836334.1 | 1 - 2 | 264 | 64 | |
| XM_004836279.3 | XP_004836336.1 | 1 - 2 | 264 | 64 | ||
More Information
Set Membership: |
|

Prelid1
Genomic Map