Assay Details
Gene Symbol: Cd99 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004867320.3 | XP_004867377.1 | 3 - 4 | 282 | 59 | |
XM_004867321.3 | XP_004867378.1 | 3 - 4 | 282 | 59 | ||
XM_021246367.1 | XP_021102026.1 | 3 - 4 | 283 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
