Assay Details
| Gene Symbol: Tmem45b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004862629.2 | XP_004862686.1 | 3 - 4 | 304 | 64 | |
| XM_004862630.3 | XP_004862687.1 | 2 - 3 | 262 | 64 | ||
| XM_021242822.1 | XP_021098481.1 | 3 - 4 | 278 | 64 | ||
More Information
Set Membership: |
|

Tmem45b
Genomic Map