Assay Details
| Gene Symbol: Hsdl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004842772.2 | XP_004842829.1 | 3 - 4 | 392 | 75 | |
| XM_004842773.2 | XP_004842830.1 | 2 - 3 | 410 | 75 | ||
| XM_004842774.2 | XP_004842831.1 | 2 - 3 | 324 | 75 | ||
| XM_013071473.2 | XP_012926927.1 | 3 - 4 | 470 | 75 | ||
More Information
Set Membership: |
|

Hsdl1
Genomic Map