Assay Details
| Gene Symbol: Rtn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004837454.2 | XP_004837511.1 | 5 - 6 | 2645 | 57 | |
| XM_004837455.2 | XP_004837512.1 | 3 - 4 | 567 | 57 | ||
| XM_021248044.1 | XP_021103703.1 | 5 - 6 | 2281 | 57 | ||
More Information
Set Membership: |
|

Rtn1
Genomic Map