Assay Details
| Gene Symbol: Glra3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004852950.2 | XP_004853007.2 | 9 - 10 | 1438 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Glra3| Gene Symbol: Glra3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004852950.2 | XP_004853007.2 | 9 - 10 | 1438 | 75 | |
Set Membership: |
|