Assay Details
| Gene Symbol: Prelid2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854341.2 | XP_004854398.1 | 5 - 6 | 608 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prelid2| Gene Symbol: Prelid2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854341.2 | XP_004854398.1 | 5 - 6 | 608 | 73 | |
Set Membership: |
|