Assay Details
| Gene Symbol: Tmub1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004860382.3 | XP_004860439.1 | 2 - 3 | 587 | 79 | |
| XM_021240974.1 | XP_021096633.1 | 2 - 3 | 663 | 79 | ||
More Information
Set Membership: |
|

Tmub1
Genomic Map