Assay Details
| Gene Symbol: Yipf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004846491.3 | XP_004846548.1 | 8 - 9 | 1057 | 70 | |
| XM_021259769.1 | XP_021115428.1 | 7 - 8 | 966 | 70 | ||
More Information
Set Membership: |
|

Yipf3
Genomic Map