Assay Details
| Gene Symbol: Ccpg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021260782.1 | XP_021116441.1 | 7 - 8 | 2263 | 109 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccpg1| Gene Symbol: Ccpg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021260782.1 | XP_021116441.1 | 7 - 8 | 2263 | 109 | |
Set Membership: |
|