Assay Details
| Gene Symbol: Cpt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004874667.2 | XP_004874724.1 | 4 - 5 | 1778 | 78 | |
| XM_004874668.2 | XP_004874725.1 | 4 - 5 | 1778 | 78 | ||
More Information
Set Membership: |
|

Cpt2
Genomic Map