Assay Details
| Gene Symbol: Fam109b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021258911.1 | XP_021114570.1 | 3 - 4 | 1022 | 64 | |
| XM_021258912.1 | XP_021114571.1 | 3 - 4 | 252 | 64 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021258911.1, XM_021258912.1 transcripts.
Set Membership: |
|

Fam109b
Genomic Map