Assay Details
| Gene Symbol: C8g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004848821.3 | XP_004848878.1 | 5 - 6 | 689 | 68 | |
| XM_013073810.2 | XP_012929264.1 | 5 - 6 | 707 | 68 | ||
| XM_013073811.2 | XP_012929265.1 | 5 - 6 | 653 | 68 | ||
| XM_021261641.1 | XP_021117300.1 | 5 - 6 | 570 | 68 | ||
| XM_021261642.1 | XP_021117301.1 | 5 - 6 | 611 | 68 | ||
More Information
Set Membership: |
|

C8g
Genomic Map