Assay Details
| Gene Symbol: Patz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843482.2 | XP_004843539.1 | 2 - 3 | 1726 | 70 | |
| XM_004843483.2 | XP_004843540.1 | 2 - 3 | 1612 | 70 | ||
| XM_004843485.2 | XP_004843542.1 | 2 - 3 | 1621 | 70 | ||
More Information
Set Membership: |
|

Patz1
Genomic Map