Assay Details
| Gene Symbol: Rab3ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845053.2 | XP_004845110.1 | 6 - 7 | 1077 | 87 | |
| XM_004845055.3 | XP_004845112.1 | 3 - 4 | 442 | 87 | ||
| XM_021258531.1 | XP_021114190.1 | 7 - 8 | 1299 | 87 | ||
More Information
Set Membership: |
|

Rab3ip
Genomic Map