Assay Details
| Gene Symbol: Il11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021245747.1 | XP_021101406.1 | 1 - 2 | 221 | 102 | |
| XM_021245748.1 | XP_021101407.1 | 1 - 2 | 221 | 102 | ||
| XM_021245749.1 | XP_021101408.1 | 1 - 2 | 221 | 102 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021245747.1, XM_021245748.1, XM_021245749.1 transcripts.
Set Membership: |
|

Il11
Genomic Map